missing translation for 'onlineSavingsMsg'
Learn More

Cytidine 5'-Monophosphate Disodium Salt Hydrate 99.0+%, TCI America™

$38.51 - $53.90

Chemical Identifiers

CAS 6757-06-8
Molecular Formula C9H12N3Na2O8P
Molecular Weight (g/mol) 367.16
MDL Number MFCD00150830
InChI Key INTPYBRGLGSMRA-IJRYJGAHNA-L
Synonym cytidine 5'-monophosphate disodium salt, disodium cmp, cmp disodium salt, cytidine-5'-monophosphate disodium salt, disodium 5'-cmp, 5'-cmp disodium salt, disodium 5'-o-phosphonatocytidine, 5'-cytidylic acid, disodium salt, disodium cytidine 5'-monophosphate, cytidine monophosphate disodium salt
PubChem CID 94332
IUPAC Name disodium [(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C05241G
View Documents
TCI America
C05241G
1 g
Each for $38.51
Only null left
 
C05245G
View Documents
TCI America
C05245G
5 g
Each for $53.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6757-06-8
367.16
INTPYBRGLGSMRA-IJRYJGAHNA-L
94332
[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O
C9H12N3Na2O8P
MFCD00150830
cytidine 5'-monophosphate disodium salt, disodium cmp, cmp disodium salt, cytidine-5'-monophosphate disodium salt, disodium 5'-cmp, 5'-cmp disodium salt, disodium 5'-o-phosphonatocytidine, 5'-cytidylic acid, disodium salt, disodium cytidine 5'-monophosphate, cytidine monophosphate disodium salt
disodium [(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate

Specifications

6757-06-8
C9H12N3Na2O8P
1 g
INTPYBRGLGSMRA-IJRYJGAHNA-L
disodium [(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
94332
≥99.0% (HPLC)
Cytidine 5′-Monophosphate Disodium Salt Hydrate
White
MFCD00150830
cytidine 5'-monophosphate disodium salt, disodium cmp, cmp disodium salt, cytidine-5'-monophosphate disodium salt, disodium 5'-cmp, 5'-cmp disodium salt, disodium 5'-o-phosphonatocytidine, 5'-cytidylic acid, disodium salt, disodium cytidine 5'-monophosphate, cytidine monophosphate disodium salt
[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O
367.16
367.16
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0282

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'