missing translation for 'onlineSavingsMsg'
Learn More

Cytidine 5'-Monophosphate Disodium Salt Hydrate 99.0+%, TCI America™

Catalog No. C05241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05241G 1 g
C05245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C05241G Supplier TCI America Supplier No. C05241G

Chemical Identifiers

CAS 6757-06-8
Molecular Formula C9H12N3Na2O8P
Molecular Weight (g/mol) 367.16
MDL Number MFCD00150830
InChI Key INTPYBRGLGSMRA-IJRYJGAHNA-L
Synonym cytidine 5'-monophosphate disodium salt, disodium cmp, cmp disodium salt, cytidine-5'-monophosphate disodium salt, disodium 5'-cmp, 5'-cmp disodium salt, disodium 5'-o-phosphonatocytidine, 5'-cytidylic acid, disodium salt, disodium cytidine 5'-monophosphate, cytidine monophosphate disodium salt
PubChem CID 94332
IUPAC Name disodium [(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O

Specifications

Color White
Quantity 1 g
Formula Weight 367.16
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cytidine 5′-Monophosphate Disodium Salt Hydrate
EINECSNumber (8)-0282
TSCA No
Recommended Storage Refrigerator