missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Cytidine-5'-monophosphate disodium salt, 99+%
CAS: 6757-06-8 | C9H12N3Na2O8P | 367.16 g/mol
Supplier: Thermo Scientific Chemicals J6337614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cytidine-5'-monophosphate disodium salt | |
| White | |
| MFCD00150830 | |
| cytidine 5'-monophosphate disodium salt, disodium cmp, cmp disodium salt, cytidine-5'-monophosphate disodium salt, disodium 5'-cmp, 5'-cmp disodium salt, disodium 5'-o-phosphonatocytidine, 5'-cytidylic acid, disodium salt, disodium cytidine 5'-monophosphate, cytidine monophosphate disodium salt | |
| [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O | |
| 367.16 | |
| 367.16 | |
| Crystalline Powder |
| 6757-06-8 | |
| C9H12N3Na2O8P | |
| 25 g | |
| INTPYBRGLGSMRA-IJRYJGAHNA-L | |
| disodium;[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate | |
| 94332 | |
| ≥99% |
Chemical Identifiers
| 6757-06-8 | |
| 367.16 | |
| INTPYBRGLGSMRA-IJRYJGAHNA-L | |
| 94332 | |
| [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O |
| C9H12N3Na2O8P | |
| MFCD00150830 | |
| cytidine 5'-monophosphate disodium salt, disodium cmp, cmp disodium salt, cytidine-5'-monophosphate disodium salt, disodium 5'-cmp, 5'-cmp disodium salt, disodium 5'-o-phosphonatocytidine, 5'-cytidylic acid, disodium salt, disodium cytidine 5'-monophosphate, cytidine monophosphate disodium salt | |
| disodium;[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only