missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytidine 5'-Diphosphate Trisodium Salt Hydrate 95.0+%, TCI America™
$39.14 - $497.70
Chemical Identifiers
| CAS | 34393-59-4 |
|---|---|
| Molecular Formula | C9H12N3Na3O11P2 |
| Molecular Weight (g/mol) | 469.12 |
| MDL Number | MFCD00065201 |
| InChI Key | JENUKVZGXGULDX-LLWADOMFSA-K |
| Synonym | cytidine 5'-diphosphate trisodium salt |
| PubChem CID | 131674171 |
| IUPAC Name | trisodium ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate |
| SMILES | [Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C052310MG
|
TCI America
C052310MG |
10 mg |
Each for $39.14
|
|
|||||
|
C0523100MG
|
TCI America
C0523100MG |
100 mg |
Each for $98.79
|
|
|||||
|
C05231G
|
TCI America
C05231G |
1 g |
Each for $497.70
|
|
|||||
Chemical Identifiers
| 34393-59-4 | |
| 469.12 | |
| JENUKVZGXGULDX-LLWADOMFSA-K | |
| 131674171 | |
| [Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O |
| C9H12N3Na3O11P2 | |
| MFCD00065201 | |
| cytidine 5'-diphosphate trisodium salt | |
| trisodium ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate |
Specifications
| 34393-59-4 | |
| C9H12N3Na3O11P2 | |
| 10 mg | |
| JENUKVZGXGULDX-LLWADOMFSA-K | |
| trisodium ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate | |
| 131674171 | |
| ≥95.0% (HPLC,T) | |
| Cytidine 5′-Diphosphate Trisodium Salt Hydrate |
| White | |
| MFCD00065201 | |
| cytidine 5'-diphosphate trisodium salt | |
| [Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O | |
| 469.12 | |
| 469.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator