missing translation for 'onlineSavingsMsg'
Learn More

Cytidine 5'-Diphosphate Trisodium Salt Hydrate 95.0+%, TCI America™

Supplier:  TCI America C05231G

 View more versions of this product

Catalog No. C05231G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cytidine 5′-Diphosphate Trisodium Salt Hydrate
White
MFCD00065201
cytidine 5'-diphosphate trisodium salt
[Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O
469.12
469.12
Crystalline Powder
34393-59-4
C9H12N3Na3O11P2
1 g
JENUKVZGXGULDX-LLWADOMFSA-K
trisodium ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
131674171
≥95.0% (HPLC,T)

Chemical Identifiers

34393-59-4
469.12
JENUKVZGXGULDX-LLWADOMFSA-K
131674171
[Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O
C9H12N3Na3O11P2
MFCD00065201
cytidine 5'-diphosphate trisodium salt
trisodium ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions