missing translation for 'onlineSavingsMsg'
Learn More

Cytarabine 98.0+%, TCI America™

Catalog No. p-7125249
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20351G 1 g
C20355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20351G Supplier TCI America Supplier No. C20351G
Only null left

Chemical Identifiers

CAS 147-94-4
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.219
InChI Key UHDGCWIWMRVCDJ-CCXZUQQUSA-N
Synonym cytarabine, cytosine arabinoside, ara-c, arabinocytidine, cytosar, arabinosylcytosine, aracytidine, cytarabin, cytarabinoside, tarabine
PubChem CID 6253
ChEBI CHEBI:28680
IUPAC Name 4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O

Specifications

Melting Point 214°C
Color White
Quantity 1 g
Formula Weight 243.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cytarabine
RTECSNumber HA5425000
TSCA No