missing translation for 'onlineSavingsMsg'
Learn More

Cytarabine, 98%

Catalog No. p-4813446
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449561000 100 mg Glass bottle
AC449560010 1 g Glass bottle
AC449560050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC449561000 Supplier Thermo Scientific Chemicals Supplier No. 449561000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cytarabine, CAS # 147-94-4, is an inhibitor of DNA synthesis that is used as an anticancer and antiviral agent. | CAS: 147-94-4 | C9H13N3O5 | 243.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cytarabine is a nucleoside analog that acts as an antileukemic agent
  • Cytarabine becomes cytotoxic following conversion to its triphosphorylated form, which can be incorporated into DNA primers by DNA polymerase alpha
  • Can induce S phase cell cycle arrest and apoptosis in leukemic and neuronal cells

Applications

  • Has been used to study chemotherapeutic regimens in laboratory studies of leukemia
  • Useful for laboratory studies of cytarabine resistance mechanisms
  • At nanomolar concentrations, cytarabine has been seen to induce mitochondrial apoptosis in leukemic cells through upregulation of multiple Bcl-2 family members
  • It can induce neuronal cell death via inducing γ-H2AX phosphorylation in histones, in the absence of double-stranded DNA breaks

Chemical Identifiers

CAS 147-94-4
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
InChI Key UHDGCWIWMRVCDJ-CCXZUQQUSA-N
Synonym cytarabine, cytosine arabinoside, ara-c, arabinocytidine, cytosar, arabinosylcytosine, aracytidine, cytarabin, cytarabinoside, tarabine
PubChem CID 6253
ChEBI CHEBI:28680
IUPAC Name 4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O

Specifications

Melting Point 220.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 152781
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dmso, ethanol
Formula Weight 243.22
Percent Purity 98%
Chemical Name or Material Cytarabine
Hazard Category Serious eye damage/eye irritation (category 2), Germ cell mutagenicity (category 2), Reproductive toxicity (category 2), Skin sensitisation (category 1)
Hazard Statement Causes serious eye irritation, Suspected of causing genetic defects, Suspected of damaging fertility or the unborn child, May cause an allergic skin reaction
EINECSNumber 205-705-9

RUO – Research Use Only