missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Cystatin C (CST3) ELISA Kit

ELISA

$884.00

Specifications

Accession Number P01034
Assay Range 0.313 to 20 ng/mL
Assay Sensitivity 300 pg/mL
Calibration Range 0.3 ng/mL to 20 ng/mL
Assay Solid-phase sandwich ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EHCST3
Encompass
View Documents Promotion Details
Invitrogen™
EHCST3
96 Tests
Pack for $884.00
Only null left
 
Description

Description

The Human Cystatin C (CST3) ELISA quantitates Hu Cystatin C in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Cystatin C. Principle of the method The Human Cystatin C solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Cystatin C, a member of the type 2 cystatin superfamily, is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsins), and pathogenic. Cystatin C is a 13 kDa secreted protein which is found in high concentrations in biological fluids and is thought to be expressed by all human nucleated cells. Changes in Cystatin C have been documented in a number of cardiovascular diseases, including atherosclerosis, amyloid angiopathy and myocardial infarction. Its low molecular weight also means that Cystatin C can be used as a marker of renal failure.
Specifications

Specifications

P01034
300 pg/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
Human
Colorimetric Microplate Reader
ARMD11,HEL-S-2
<10
HRP
Extracellular Matrix & Cell Adhesion
2°C to 8°C
Cystatin C
4 hrs 45 mins
0.313 to 20 ng/mL
0.3 ng/mL to 20 ng/mL
HRP
Cell Structure & Organelle Proteins
Species Reactivity Human Specificity Detects human Cystatin C in ELISAs. In sandwich ELISA assay, no cross reactivity with recombinant human Cystatin A, B, D, E/M, F, S, SA, SN, X/Z/P, or recombinant mouse Cystatin C is observed.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
1471
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 1 μL, Serum 1 μL, Supernatant 100 μL
ARMD11
1 hrs 20 mins
Videos
SDS
Documents

Documents