Learn More
Invitrogen™ Human Cystatin C (CST3) ELISA Kit
ELISA
Supplier: Invitrogen™ EHCST3
Description
The Human Cystatin C (CST3) ELISA quantitates Hu Cystatin C in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Cystatin C. Principle of the method The Human Cystatin C solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Cystatin C, a member of the type 2 cystatin superfamily, is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsins), and pathogenic. Cystatin C is a 13 kDa secreted protein which is found in high concentrations in biological fluids and is thought to be expressed by all human nucleated cells. Changes in Cystatin C have been documented in a number of cardiovascular diseases, including atherosclerosis, amyloid angiopathy and myocardial infarction. Its low molecular weight also means that Cystatin C can be used as a marker of renal failure.Chemical Identifiers
| P01034 | |
| 300 pg/mL | |
| HRP | |
| Cell Structure & Organelle Proteins | |
| Species Reactivity Human Specificity Detects human Cystatin C in ELISAs. In sandwich ELISA assay, no cross reactivity with recombinant human Cystatin A, B, D, E/M, F, S, SA, SN, X/Z/P, or recombinant mouse Cystatin C is observed. | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 1471 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| Plasma 1 μL, Serum 1 μL, Supernatant 100 μL | |
| ARMD11 | |
| 1 hrs 20 mins | |
| Solid-phase sandwich ELISA |
| 0.313 to 20 ng/mL | |
| 0.3 ng/mL to 20 ng/mL | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| Human | |
| Colorimetric Microplate Reader | |
| ARMD11,HEL-S-2 | |
| <10 | |
| HRP | |
| Extracellular Matrix & Cell Adhesion | |
| 2°C to 8°C | |
| Cystatin C | |
| 4 hrs 45 mins |
Specifications
| P01034 | |
| 300 pg/mL | |
| Solid-phase sandwich ELISA | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| 96 Assays | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 1471 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Extracellular Matrix & Cell Adhesion | |
| 2°C to 8°C | |
| Cystatin C | |
| 4 hrs 45 mins |
| 0.313 to 20 ng/mL | |
| 0.3 ng/mL to 20 ng/mL | |
| HRP | |
| Cell Structure & Organelle Proteins | |
| Species Reactivity Human Specificity Detects human Cystatin C in ELISAs. In sandwich ELISA assay, no cross reactivity with recombinant human Cystatin A, B, D, E/M, F, S, SA, SN, X/Z/P, or recombinant mouse Cystatin C is observed. | |
| Human | |
| Colorimetric Microplate Reader | |
| ARMD11,HEL-S-2 | |
| <10 | |
| HRP | |
| RUO | |
| Plasma 1 μL, Serum 1 μL, Supernatant 100 μL | |
| ARMD11 | |
| 1 hrs 20 mins |