missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cyromazine

Catalog No. AC467530050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467530050 5 g
AC467530250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467530050 Supplier Thermo Scientific Chemicals Supplier No. 467530050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66215-27-8
Molecular Formula C6H10N6

Specifications

Quantity 5 g
Melting Point 223.0°C to 227.0°C
Flash Point 100°C
Formula Weight 166.16
InChI Key LVQDKIWDGQRHTE-UHFFFAOYSA-N
SMILES NC1=NC(NC2CC2)=NC(N)=N1
IUPAC Name N2-cyclopropyl-1,3,5-triazine-2,4,6-triamine
Molecular Weight (g/mol) 166.19
Chemical Name or Material Cyromazine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 266-257-8

RUO – Research Use Only