missing translation for 'onlineSavingsMsg'
Learn More

Cyproterone Acetate 98.0+%, TCI America™

Supplier:  TCI America C29421G

Quantity 1 g

 View more versions of this product

Catalog No. C29421G


Only null left

Description

Description

Specifications

Chemical Identifiers

427-51-0
416.942
UWFYSQMTEOIJJG-FDTZYFLXSA-N
9880
CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C
C24H29ClO4
MFCD00864671
cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385
CHEBI:50743

Specifications

Cyproterone Acetate
209°C
C24H29ClO4
MFCD00864671
UWFYSQMTEOIJJG-FDTZYFLXSA-N
416.942
CHEBI:50743
≥98.0% (HPLC)
427-51-0
White
1 g
cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385
CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C
9880
416.94
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : GZ2230000

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions