accessibility menu, dialog, popup

UserName

CAS RN 427-51-0

IUPAC Name: (1S,2S,3S,5R,11R,12S,15R,16S)-15-acetyl-9-chloro-2,16-dimethyl-6-oxopentacyclo[9.7.0.0²,⁸.0³,⁵.0¹²,¹⁶]octadeca-7,9-dien-15-yl acetate
Synonyms: cyproterone acetate cyproterone preparation (product) cyproterone 17α-acetate cyproteron novo-cyproterone cyproteron acetate cyproteroni acetas cyproteron-R acetate ciproterona servycal androcur
Molecular Weight (g/mol): 416.94
Molecular Formula: C24H29ClO4
InChi Key: UWFYSQMTEOIJJG-FDTZYFLXSA-N
SMILES: CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Cl)C4=CC(=O)[C@@H]5C[C@@H]5[C@]4(C)[C@H]3CC[C@]12C)C(C)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)