missing translation for 'onlineSavingsMsg'
Learn More

Cyproterone Acetate 98.0+%, TCI America™

Catalog No. p-7125736
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2942200MG 200 mg
C29421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2942200MG Supplier TCI America Supplier No. C2942200MG

Chemical Identifiers

CAS 427-51-0
Molecular Formula C24H29ClO4
Molecular Weight (g/mol) 416.942
MDL Number MFCD00864671
InChI Key UWFYSQMTEOIJJG-FDTZYFLXSA-N
Synonym cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385
PubChem CID 9880
ChEBI CHEBI:50743
SMILES CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C

Specifications

Melting Point 209°C
Color White
Quantity 200 mg
Formula Weight 416.94
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cyproterone Acetate
RTECSNumber GZ2230000
TSCA No
Recommended Storage Freezer