missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CYM 50308

Catalog No. 467810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
467810 10 mg
467850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 467810 Supplier Tocris Bioscience™ Supplier No. 4678/10
Only null left

Potent and selective S1P4 agonist

Potent and selective sphingosine-1-phosphate receptor 4 (S1P4) agonist (EC50 values are 56 nM and 2100 nM for S1P4 and S1P5 receptors, respectively). Displays no activity at S1P1, S1P2 and S1P3 receptors at concentrations up to 25 μM.

Chemical Identifiers

CAS 1345858-76-5
Molecular Formula C20H21F2N3O2S

Specifications

Quantity 10 mg
Formula Weight 405.46
Purity >98%
Molecular Weight (g/mol) 405.464
Chemical Name or Material CYM 50308
Recommended Storage Store at +4°C