missing translation for 'onlineSavingsMsg'
Learn More

Cyhalofop Butyl 97.0+%, TCI America™

Catalog No. C24171G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24171G 1 g
C241725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24171G Supplier TCI America Supplier No. C24171G
Only null left

Chemical Identifiers

CAS 122008-85-9
Molecular Formula C20H20FNO4
Molecular Weight (g/mol) 357.381
MDL Number MFCD01631151
InChI Key TYIYMOAHACZAMQ-CQSZACIVSA-N
Synonym Butyl (R)-2-[4-(4-Cyano-2-fluorophenoxy)phenoxy]propionate, (R)-2-[4-(4-Cyano-2-fluorophenoxy)phenoxy]propionic Acid Butyl Ester
PubChem CID 180089
ChEBI CHEBI:81863
IUPAC Name butyl (2R)-2-[4-(4-cyano-2-fluorophenoxy)phenoxy]propanoate
SMILES CCCCOC(=O)C(C)OC1=CC=C(C=C1)OC2=C(C=C(C=C2)C#N)F

Specifications

Melting Point 49°C
Color White
Quantity 1 g
UN Number 3077
Formula Weight 357.38
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Cyhalofop Butyl
TSCA No
Recommended Storage Refrigerator