accessibility menu, dialog, popup

UserName

H3COO(R)CH3OOFN

CAS RN 122008-85-9

IUPAC Name: butyl (2R)-2-[4-(4-cyano-2-fluorophenoxy)phenoxy]propanoate
Synonyms: butyl (2R)-2-[4-(4-cyano-2-fluorophenoxy)phenoxy]propanoate
Molecular Weight (g/mol): 357.38
Molecular Formula: C20H20FNO4
InChi Key: TYIYMOAHACZAMQ-CQSZACIVSA-N
SMILES: CCCCOC(=O)[C@@H](C)OC1=CC=C(OC2=CC=C(C=C2F)C#N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results