Learn More
Cyclopropyltriphenylphosphonium bromide, 98%
CAS: 14114-05-7 | C21H20BrP | 383.27 g/mol
$118.44 - $1271.01
Chemical Identifiers
| CAS | 14114-05-7 |
|---|---|
| Molecular Formula | C21H20BrP |
| Molecular Weight (g/mol) | 383.27 |
| MDL Number | MFCD00011872 |
| InChI Key | XMPWFKHMCNRJCL-UHFFFAOYSA-M |
| Synonym | cyclopropyltriphenylphosphonium bromide, phosphonium, cyclopropyltriphenyl-, bromide, cyclopropyltriphenylphosphanium bromide, cyclopropyl triphenyl phosphanium bromide, acmc-209cls, cyclopropyltriphenylphosphoniumbromide, cyclopropyl triphenylphosphonium bromide, cyclopropyl-triphenylphosphonium bromide, cyclopropyltriphenyl-phosphonium bromide, triphenyl-cyclopropylphosphonium bromide |
| PubChem CID | 2723931 |
| IUPAC Name | cyclopropyl(triphenyl)phosphanium;bromide |
| SMILES | [Br-].C1CC1[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2510206
|
Thermo Scientific Chemicals
B2510206 |
5 g |
Each for $118.44
|
|
|||||
|
AAB2510214
|
Thermo Scientific Chemicals
B2510214 |
25 g |
Each for $418.38
|
|
|||||
|
AAB2510222
|
Thermo Scientific Chemicals
B2510222 |
100 g |
Each for $1,271.01
|
|
|||||
Description
It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Chemical Identifiers
| 14114-05-7 | |
| 383.27 | |
| XMPWFKHMCNRJCL-UHFFFAOYSA-M | |
| 2723931 | |
| [Br-].C1CC1[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C21H20BrP | |
| MFCD00011872 | |
| cyclopropyltriphenylphosphonium bromide, phosphonium, cyclopropyltriphenyl-, bromide, cyclopropyltriphenylphosphanium bromide, cyclopropyl triphenyl phosphanium bromide, acmc-209cls, cyclopropyltriphenylphosphoniumbromide, cyclopropyl triphenylphosphonium bromide, cyclopropyl-triphenylphosphonium bromide, cyclopropyltriphenyl-phosphonium bromide, triphenyl-cyclopropylphosphonium bromide | |
| cyclopropyl(triphenyl)phosphanium;bromide |
Specifications
| 14114-05-7 | |
| C21H20BrP | |
| 5 g | |
| Hygroscopic | |
| Soluble in water. | |
| [Br-].C1CC1[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 383.27 | |
| 383.27 | |
| Cyclopropyltriphenylphosphonium bromide |
| 183°C to 186°C | |
| MFCD00011872 | |
| 6413483 | |
| cyclopropyltriphenylphosphonium bromide, phosphonium, cyclopropyltriphenyl-, bromide, cyclopropyltriphenylphosphanium bromide, cyclopropyl triphenyl phosphanium bromide, acmc-209cls, cyclopropyltriphenylphosphoniumbromide, cyclopropyl triphenylphosphonium bromide, cyclopropyl-triphenylphosphonium bromide, cyclopropyltriphenyl-phosphonium bromide, triphenyl-cyclopropylphosphonium bromide | |
| XMPWFKHMCNRJCL-UHFFFAOYSA-M | |
| cyclopropyl(triphenyl)phosphanium;bromide | |
| 2723931 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
H312+H332
EINECSNumber : 237-970-1
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only