missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyltriphenylphosphonium bromide, 98%

CAS: 14114-05-7 | C21H20BrP | 383.27 g/mol

Supplier:  Thermo Scientific Chemicals B2510214

 View more versions of this product

Catalog No. AAB2510214

Only null left
Explore available promotions

Description

Description

It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Specifications

Specifications

Cyclopropyltriphenylphosphonium bromide
183°C to 186°C
MFCD00011872
6413483
cyclopropyltriphenylphosphonium bromide, phosphonium, cyclopropyltriphenyl-, bromide, cyclopropyltriphenylphosphanium bromide, cyclopropyl triphenyl phosphanium bromide, acmc-209cls, cyclopropyltriphenylphosphoniumbromide, cyclopropyl triphenylphosphonium bromide, cyclopropyl-triphenylphosphonium bromide, cyclopropyltriphenyl-phosphonium bromide, triphenyl-cyclopropylphosphonium bromide
XMPWFKHMCNRJCL-UHFFFAOYSA-M
cyclopropyl(triphenyl)phosphanium;bromide
2723931
98%
14114-05-7
C21H20BrP
25 g
Hygroscopic
Soluble in water.
[Br-].C1CC1[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
383.27
383.27

Chemical Identifiers

14114-05-7
383.27
XMPWFKHMCNRJCL-UHFFFAOYSA-M
2723931
[Br-].C1CC1[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C21H20BrP
MFCD00011872
cyclopropyltriphenylphosphonium bromide, phosphonium, cyclopropyltriphenyl-, bromide, cyclopropyltriphenylphosphanium bromide, cyclopropyl triphenyl phosphanium bromide, acmc-209cls, cyclopropyltriphenylphosphoniumbromide, cyclopropyl triphenylphosphonium bromide, cyclopropyl-triphenylphosphonium bromide, cyclopropyltriphenyl-phosphonium bromide, triphenyl-cyclopropylphosphonium bromide
cyclopropyl(triphenyl)phosphanium;bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c

H312+H332

EINECSNumber : 237-970-1

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only