missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropanecarboxaldehyde 98.0+%, TCI America™
Supplier: TCI America C17075G
Specifications
| Cyclopropanecarboxaldehyde | |
| Yellow | |
| C4H6O | |
| 5 g | |
| cyclopropanecarboxaldehyde, formylcyclopropane, cyclopropylmethanal, cyclopropanecarboxyldehyde, cyclopropanal, cyclopropane aldehyde, cyclopropancarbaldehyd, cyclopropancarbaldehyde, cyclopropancarboxaldehyde, cyclopropanecarboaldehyde | |
| C1CC1C=O | |
| 70.091 | |
| 70.09 | |
| Liquid |
| 1489-69-6 | |
| 102°C | |
| MFCD00012261 | |
| 2924 | |
| JMYVMOUINOAAPA-UHFFFAOYSA-N | |
| cyclopropanecarbaldehyde | |
| 123114 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1489-69-6 | |
| 70.091 | |
| JMYVMOUINOAAPA-UHFFFAOYSA-N | |
| 123114 | |
| C1CC1C=O |
| C4H6O | |
| MFCD00012261 | |
| cyclopropanecarboxaldehyde, formylcyclopropane, cyclopropylmethanal, cyclopropanecarboxyldehyde, cyclopropanal, cyclopropane aldehyde, cyclopropancarbaldehyd, cyclopropancarbaldehyde, cyclopropancarboxaldehyde, cyclopropanecarboaldehyde | |
| cyclopropanecarbaldehyde |
Safety and Handling
EINECSNumber : (3)-4433
TSCA : Yes
Recommended Storage : Refrigerator