missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxaldehyde 98.0+%, TCI America™

Catalog No. p-7125132
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
C17071G 1 g
C17075G 5 g
2 options

Catalog No. C17071G

Supplier: TCI America C17071G

Only null left

Chemical Identifiers

CAS 1489-69-6
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00012261
InChI Key JMYVMOUINOAAPA-UHFFFAOYSA-N
Synonym cyclopropanecarboxaldehyde, formylcyclopropane, cyclopropylmethanal, cyclopropanecarboxyldehyde, cyclopropanal, cyclopropane aldehyde, cyclopropancarbaldehyd, cyclopropancarbaldehyde, cyclopropancarboxaldehyde, cyclopropanecarboaldehyde
PubChem CID 123114
IUPAC Name cyclopropanecarbaldehyde
SMILES C1CC1C=O

Specifications

Color Yellow
Boiling Point 102°C
Quantity 1 g
UN Number 2924
Formula Weight 70.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopropanecarboxaldehyde
EINECSNumber (3)-4433
TSCA Yes
Recommended Storage Refrigerator