missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyltriphenylphosphonium bromide, 96%

Catalog No. AAA1464709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1464709 10 g
AAA1464718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1464709 Supplier Thermo Scientific Chemicals Supplier No. A1464709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7333-52-0 | C23H24BrP | 411.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7333-52-0
Molecular Formula C23H24BrP
Molecular Weight (g/mol) 411.32
MDL Number MFCD00051880
InChI Key WZYWSVSFFTZZPE-UHFFFAOYSA-M
Synonym cyclopentyltriphenylphosphonium bromide, cyclopentyl triphenyl phosphanium bromide, phosphonium, cyclopentyltriphenyl-, bromide, cyclopentyltriphenylphosphanium bromide, cyclopentyl triphenyl phosphonium bromide, cyclopentyltriphenylphosphoniumbromide, cyclopentyl triphenylphosphonium bromide, phosphonium,cyclopentyltriphenyl-,bromide 1:1, phosphonium,cyclopentyltriphenyl-, bromide 1:1
PubChem CID 2735896
IUPAC Name cyclopentyl(triphenyl)phosphanium;bromide
SMILES [Br-].C1CCC(C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 262°C to 266°C
Quantity 10 g
Beilstein 3580193
Sensitivity Hygroscopic
Formula Weight 411.33
Percent Purity 98%
Chemical Name or Material Cyclopentyltriphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only