CAS RN 7333-52-0
IUPAC Name:
cyclopentyltriphenylphosphanium bromide
Synonyms:
cyclopentyltriphenylphosphanium bromide
Molecular Weight (g/mol):
411.32
Molecular Formula:
C23H24BrP
InChi Key:
WZYWSVSFFTZZPE-UHFFFAOYSA-M
SMILES:
[Br-].C1CCC(C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Cyclopentyltriphenylphosphonium bromide, 96%
Cyclopentyltriphenylphosphonium bromide, 96%
CAS: 7333-52-0 Molecular Formula: C23H24BrP Molecular Weight (g/mol): 411.32 MDL Number: MFCD00051880 InChI Key: WZYWSVSFFTZZPE-UHFFFAOYSA-M Synonym: cyclopentyltriphenylphosphonium bromide,cyclopentyl triphenyl phosphanium bromide,phosphonium, cyclopentyltriphenyl-, bromide,cyclopentyltriphenylphosphanium bromide,cyclopentyl triphenyl phosphonium bromide,cyclopentyltriphenylphosphoniumbromide,cyclopentyl triphenylphosphonium bromide,phosphonium,cyclopentyltriphenyl-,bromide 1:1,phosphonium,cyclopentyltriphenyl-, bromide 1:1 PubChem CID: 2735896 IUPAC Name: cyclopentyl(triphenyl)phosphanium;bromide SMILES: [Br-].C1CCC(C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1