missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentanecarbonitrile 98.0+%, TCI America™
Supplier: TCI America C316725ML
Specifications
| Cyclopentanecarbonitrile | |
| Colorless | |
| C6H9N | |
| 25 mL | |
| cyanocyclopentane, cyclopentyl cyanide, cyclopentane carbonitrile, cyclopentanenitrile, cyclopentyl nitrile, 1-cyanocyclopentane, cyclopentankohlenitrile, acmc-1aqy6, ksc238k0j, cyclopentanecarbonitrile | |
| C1CCC(C1)C#N | |
| 95.15 | |
| 95.15 | |
| Liquid |
| 4254-02-8 | |
| 168°C | |
| MFCD00013741 | |
| 1992 | |
| SVPZJHKVRMRREG-UHFFFAOYSA-N | |
| cyclopentanecarbonitrile | |
| 77935 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 4254-02-8 | |
| 95.15 | |
| SVPZJHKVRMRREG-UHFFFAOYSA-N | |
| 77935 | |
| C1CCC(C1)C#N |
| C6H9N | |
| MFCD00013741 | |
| cyanocyclopentane, cyclopentyl cyanide, cyclopentane carbonitrile, cyclopentanenitrile, cyclopentyl nitrile, 1-cyanocyclopentane, cyclopentankohlenitrile, acmc-1aqy6, ksc238k0j, cyclopentanecarbonitrile | |
| cyclopentanecarbonitrile |
Safety and Handling
TSCA : No