accessibility menu, dialog, popup

UserName

CAS RN 4254-02-8

IUPAC Name: cyclopentanecarbonitrile
Synonyms: cyclopentanecarbonitrile
Molecular Weight (g/mol): 95.15
Molecular Formula: C6H9N
InChi Key: SVPZJHKVRMRREG-UHFFFAOYSA-N
SMILES: N#CC1CCCC1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)