missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarbonitrile 98.0+%, TCI America™

Catalog No. p-7125869
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31675ML 5 mL
C316725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31675ML Supplier TCI America Supplier No. C31675ML

Chemical Identifiers

CAS 4254-02-8
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.15
MDL Number MFCD00013741
InChI Key SVPZJHKVRMRREG-UHFFFAOYSA-N
Synonym cyanocyclopentane, cyclopentyl cyanide, cyclopentane carbonitrile, cyclopentanenitrile, cyclopentyl nitrile, 1-cyanocyclopentane, cyclopentankohlenitrile, acmc-1aqy6, ksc238k0j, cyclopentanecarbonitrile
PubChem CID 77935
IUPAC Name cyclopentanecarbonitrile
SMILES C1CCC(C1)C#N

Specifications

Color Colorless
Boiling Point 168°C
Quantity 5 mL
UN Number 1992
Formula Weight 95.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentanecarbonitrile