missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopenta[fg]tetracene-1,2-dione, TCI America™
Supplier: TCI America C26691G
Specifications
| Cyclopenta[fg]tetracene-1,2-dione | |
| Blue-Purple | |
| 1 g | |
| AYSSMZWWOZPRMT-UHFFFAOYSA-N | |
| 282.298 | |
| 282.30 |
| 1334510-66-5 | |
| C20H10O2 | |
| Cyclopenta[fg]naphthacene-1,2-dione | |
| C1=CC=C2C(=C1)C=C3C=C4C=CC=CC4=C5C3=C2C(=O)C5=O | |
| 91972158 | |
| Crystalline Powder |
Chemical Identifiers
| 1334510-66-5 | |
| 282.298 | |
| Cyclopenta[fg]naphthacene-1,2-dione | |
| C1=CC=C2C(=C1)C=C3C=C4C=CC=CC4=C5C3=C2C(=O)C5=O |
| C20H10O2 | |
| AYSSMZWWOZPRMT-UHFFFAOYSA-N | |
| 91972158 |
Safety and Handling
TSCA : No