missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyldiphenylphosphine Oxide 98.0+%, TCI America™

Catalog No. p-7125922
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33101G 1 g
C33105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33101G Supplier TCI America Supplier No. C33101G

Chemical Identifiers

CAS 13689-20-8
Molecular Formula C18H21OP
Molecular Weight (g/mol) 284.34
MDL Number MFCD00014301
InChI Key ICVUZKQDJNUMKC-UHFFFAOYSA-N
Synonym cyclohexyldiphenylphosphine oxide, diphenyl cyclohexyl phosphine oxide, cyclohexyldiphenylphosphineoxide, phosphine oxide, cyclohexyldiphenyl, cyclohexyl diphenyl phosphineoxide, acmc-20alcj, diphenylphosphinyl cyclohexane, cyclohexyldiphenylphosphino-1-one, cyclohexyl-phenylphosphoryl benzene, cyclohexyl phenyl phosphoryl benzene
PubChem CID 83664
IUPAC Name [cyclohexyl(phenyl)phosphoroso]benzene
SMILES O=P(C1CCCCC1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 1 g
Formula Weight 284.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexyldiphenylphosphine Oxide