accessibility menu, dialog, popup

UserName

OP

CAS RN 13689-20-8

IUPAC Name: [cyclohexyl(phenyl)phosphoroso]benzene
Synonyms: [cyclohexyl(phenyl)phosphoroso]benzene
Molecular Weight (g/mol): 284.34
Molecular Formula: C18H21OP
InChi Key: ICVUZKQDJNUMKC-UHFFFAOYSA-N
SMILES: O=P(C1CCCCC1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)