missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyldiphenylphosphine oxide, 98+%

Catalog No. AAL0069806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0069806 5 g
AAL0069803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0069806 Supplier Thermo Scientific Chemicals Supplier No. L0069806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13689-20-8 | C18H21OP | 284.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13689-20-8
Molecular Formula C18H21OP
Molecular Weight (g/mol) 284.34
MDL Number MFCD00014301
InChI Key ICVUZKQDJNUMKC-UHFFFAOYSA-N
Synonym cyclohexyldiphenylphosphine oxide, diphenyl cyclohexyl phosphine oxide, cyclohexyldiphenylphosphineoxide, phosphine oxide, cyclohexyldiphenyl, cyclohexyl diphenyl phosphineoxide, acmc-20alcj, diphenylphosphinyl cyclohexane, cyclohexyldiphenylphosphino-1-one, cyclohexyl-phenylphosphoryl benzene, cyclohexyl phenyl phosphoryl benzene
PubChem CID 83664
IUPAC Name [cyclohexyl(phenyl)phosphoryl]benzene
SMILES O=P(C1CCCCC1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 166°C to 167°C
Quantity 5 g
Beilstein 2943737
Formula Weight 284.34
Percent Purity ≥98%
Chemical Name or Material Cyclohexyldiphenylphosphine oxide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-204-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only