Learn More
Cyclohexyl methyl ketone, 95%
CAS: 823-76-7 | C8H14O | 126.199 g/mol
$86.86 - $902.07
Chemical Identifiers
| CAS | 823-76-7 |
|---|---|
| Molecular Formula | C8H14O |
| Molecular Weight (g/mol) | 126.199 |
| MDL Number | MFCD00040418 |
| InChI Key | RIFKADJTWUGDOV-UHFFFAOYSA-N |
| Synonym | 1-cyclohexylethan-1-one, cyclohexyl methyl ketone, acetylcyclohexane, ethanone, 1-cyclohexyl, cyclohexylethanone, methyl cyclohexyl ketone, 1-acetylcyclohexane, cyclohexane, acetyl, acetophenone, hexahydro, ketone, cyclohexyl methyl |
| PubChem CID | 13207 |
| IUPAC Name | 1-cyclohexylethanone |
| SMILES | CC(=O)C1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2100006
|
Thermo Scientific Chemicals
B2100006 |
5 g |
Each for $86.86
|
|
|||||
|
AAB2100014
|
Thermo Scientific Chemicals
B2100014 |
25 g |
Each for $273.62
|
|
|||||
|
AAB2100022
|
Thermo Scientific Chemicals
B2100022 |
100 g |
Each for $902.07
|
|
|||||
Description
It can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 823-76-7 | |
| 126.199 | |
| RIFKADJTWUGDOV-UHFFFAOYSA-N | |
| 13207 | |
| CC(=O)C1CCCCC1 |
| C8H14O | |
| MFCD00040418 | |
| 1-cyclohexylethan-1-one, cyclohexyl methyl ketone, acetylcyclohexane, ethanone, 1-cyclohexyl, cyclohexylethanone, methyl cyclohexyl ketone, 1-acetylcyclohexane, cyclohexane, acetyl, acetophenone, hexahydro, ketone, cyclohexyl methyl | |
| 1-cyclohexylethanone |
Specifications
| 823-76-7 | |
| 0.917 | |
| 66°C (151°F) | |
| C8H14O | |
| MFCD00040418 | |
| 507229 | |
| Not miscible or difficult to mix in water. | |
| CC(=O)C1CCCCC1 | |
| 126.199 | |
| 126.2 | |
| Liquid |
| -34°C | |
| 181°C to 183°C | |
| 0.95 | |
| 1.452 | |
| 5 g | |
| 1-cyclohexylethan-1-one, cyclohexyl methyl ketone, acetylcyclohexane, ethanone, 1-cyclohexyl, cyclohexylethanone, methyl cyclohexyl ketone, 1-acetylcyclohexane, cyclohexane, acetyl, acetophenone, hexahydro, ketone, cyclohexyl methyl | |
| RIFKADJTWUGDOV-UHFFFAOYSA-N | |
| 1-cyclohexylethanone | |
| 13207 | |
| 95% | |
| Cyclohexyl methyl ketone |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 212-517-0
RTECSNumber : KM5638850
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only