missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl methyl ketone, 95%

CAS: 823-76-7 | C8H14O | 126.199 g/mol

Supplier:  Thermo Scientific Chemicals B2100022

 View more versions of this product

Catalog No. AAB2100022


Only null left
Explore available promotions

Description

Description

It can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Cyclohexyl methyl ketone
-34°C
181°C to 183°C
0.95
1.452
100 g
1-cyclohexylethan-1-one, cyclohexyl methyl ketone, acetylcyclohexane, ethanone, 1-cyclohexyl, cyclohexylethanone, methyl cyclohexyl ketone, 1-acetylcyclohexane, cyclohexane, acetyl, acetophenone, hexahydro, ketone, cyclohexyl methyl
RIFKADJTWUGDOV-UHFFFAOYSA-N
1-cyclohexylethanone
13207
95%
823-76-7
0.917
66°C (151°F)
C8H14O
MFCD00040418
507229
Not miscible or difficult to mix in water.
CC(=O)C1CCCCC1
126.199
126.2
Liquid

Chemical Identifiers

823-76-7
126.199
RIFKADJTWUGDOV-UHFFFAOYSA-N
13207
CC(=O)C1CCCCC1
C8H14O
MFCD00040418
1-cyclohexylethan-1-one, cyclohexyl methyl ketone, acetylcyclohexane, ethanone, 1-cyclohexyl, cyclohexylethanone, methyl cyclohexyl ketone, 1-acetylcyclohexane, cyclohexane, acetyl, acetophenone, hexahydro, ketone, cyclohexyl methyl
1-cyclohexylethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 212-517-0

RTECSNumber : KM5638850

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only