missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanepropionic Acid 99.0+%, TCI America™
Chemical Identifiers
| CAS | 701-97-3 |
|---|---|
| Molecular Formula | C9H16O2 |
| Molecular Weight (g/mol) | 156.23 |
| MDL Number | MFCD00001527 |
| InChI Key | HJZLEGIHUQOJBA-UHFFFAOYSA-N |
| Synonym | cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid |
| PubChem CID | 69702 |
| IUPAC Name | 3-cyclohexylpropanoic acid |
| SMILES | OC(=O)CCC1CCCCC1 |
Chemical Identifiers
| C9H16O2 | |
| MFCD00001527 | |
| cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid | |
| 3-cyclohexylpropanoic acid |
Specifications
| 701-97-3 | |
| C9H16O2 | |
| cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid | |
| OC(=O)CCC1CCCCC1 | |
| 156.23 | |
| 156.23 | |
| Liquid |
| 268°C | |
| MFCD00001527 | |
| HJZLEGIHUQOJBA-UHFFFAOYSA-N | |
| 3-cyclohexylpropanoic acid | |
| 69702 | |
| ≥99.0% (GC,T) | |
| Cyclohexanepropionic Acid |
Safety and Handling
EINECSNumber : (3)-2422
TSCA : Yes