missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanepropionic Acid 99.0+%, TCI America™

Catalog No. C048425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C048425G 25 g
C0484100G 100 g
C0484500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C048425G Supplier TCI America Supplier No. C048425G

Chemical Identifiers

CAS 701-97-3
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD00001527
InChI Key HJZLEGIHUQOJBA-UHFFFAOYSA-N
Synonym cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid
PubChem CID 69702
IUPAC Name 3-cyclohexylpropanoic acid
SMILES OC(=O)CCC1CCCCC1

Specifications

Boiling Point 268°C
Quantity 25 g
Formula Weight 156.23
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclohexanepropionic Acid
EINECSNumber (3)-2422
TSCA Yes