accessibility menu, dialog, popup

UserName

HOO

CAS RN 701-97-3

IUPAC Name: 3-cyclohexylpropanoic acid
Synonyms: cyclohexane propionic acid
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: HJZLEGIHUQOJBA-UHFFFAOYSA-N
SMILES: OC(=O)CCC1CCCCC1
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)