missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclododecanol 98.0+%, TCI America™
Supplier: TCI America C046125G
Specifications
| Cyclododecanol | |
| 77°C | |
| 104°C | |
| MFCD00003717 | |
| 3077 | |
| SFVWPXMPRCIVOK-UHFFFAOYSA-N | |
| cyclododecanol | |
| 15595 | |
| ≥98.0% (GC) |
| 1724-39-6 | |
| White | |
| C12H24O | |
| 25 g | |
| unii-0z645y4jts, dsstox_cid_7429, dsstox_rid_78451, dsstox_gsid_27429, acmc-1bprz | |
| C1CCCCCC(CCCCC1)O | |
| 184.323 | |
| 184.32 | |
| Crystalline Powder |
Chemical Identifiers
| 1724-39-6 | |
| 184.323 | |
| SFVWPXMPRCIVOK-UHFFFAOYSA-N | |
| 15595 | |
| C1CCCCCC(CCCCC1)O |
| C12H24O | |
| MFCD00003717 | |
| unii-0z645y4jts, dsstox_cid_7429, dsstox_rid_78451, dsstox_gsid_27429, acmc-1bprz | |
| cyclododecanol |
Safety and Handling
EINECSNumber : (3)-2299
TSCA : Yes