accessibility menu, dialog, popup

UserName

OH

CAS RN 1724-39-6

IUPAC Name: cyclododecanol
Synonyms: cyclododecanol
Molecular Weight (g/mol): 184.32
Molecular Formula: C12H24O
InChi Key: SFVWPXMPRCIVOK-UHFFFAOYSA-N
SMILES: OC1CCCCCCCCCCC1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)