missing translation for 'onlineSavingsMsg'
Learn More

Cyclododecanol 98.0+%, TCI America™

Catalog No. p-7124438
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C046125G 25 g
C0461500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C046125G Supplier TCI America Supplier No. C046125G
Only null left

Chemical Identifiers

CAS 1724-39-6
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.323
MDL Number MFCD00003717
InChI Key SFVWPXMPRCIVOK-UHFFFAOYSA-N
Synonym unii-0z645y4jts, dsstox_cid_7429, dsstox_rid_78451, dsstox_gsid_27429, acmc-1bprz
PubChem CID 15595
IUPAC Name cyclododecanol
SMILES C1CCCCCC(CCCCC1)O

Specifications

Melting Point 77°C
Color White
Boiling Point 104°C
Quantity 25 g
UN Number 3077
Formula Weight 184.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclododecanol
EINECSNumber (3)-2299
TSCA Yes