missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclobutanemethanol 98.0+%, TCI America™
Supplier: TCI America C206125G
Specifications
| Cyclobutanemethanol | |
| Colorless | |
| C5H10O | |
| 25 g | |
| cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465 | |
| C1CC(C1)CO | |
| 86.134 | |
| 86.13 | |
| Liquid |
| 4415-82-1 | |
| 144°C | |
| MFCD00001330 | |
| 1987 | |
| WPOPOPFNZYPKAV-UHFFFAOYSA-N | |
| cyclobutylmethanol | |
| 78130 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 4415-82-1 | |
| 86.134 | |
| WPOPOPFNZYPKAV-UHFFFAOYSA-N | |
| 78130 | |
| C1CC(C1)CO |
| C5H10O | |
| MFCD00001330 | |
| cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465 | |
| cyclobutylmethanol |
Safety and Handling
TSCA : No