accessibility menu, dialog, popup

UserName

HO

CAS RN 4415-82-1

IUPAC Name: cyclobutylmethanol
Synonyms: cyclobutylmethanol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: WPOPOPFNZYPKAV-UHFFFAOYSA-N
SMILES: OCC1CCC1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)