missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclobutanemethanol 98.0+%, TCI America™
$190.59 - $558.30
Chemical Identifiers
| CAS | 4415-82-1 |
|---|---|
| Molecular Formula | C5H10O |
| Molecular Weight (g/mol) | 86.134 |
| MDL Number | MFCD00001330 |
| InChI Key | WPOPOPFNZYPKAV-UHFFFAOYSA-N |
| Synonym | cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465 |
| PubChem CID | 78130 |
| IUPAC Name | cyclobutylmethanol |
| SMILES | C1CC(C1)CO |
Chemical Identifiers
| 4415-82-1 | |
| 86.134 | |
| WPOPOPFNZYPKAV-UHFFFAOYSA-N | |
| 78130 | |
| C1CC(C1)CO |
| C5H10O | |
| MFCD00001330 | |
| cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465 | |
| cyclobutylmethanol |
Specifications
| 4415-82-1 | |
| 144°C | |
| MFCD00001330 | |
| 1987 | |
| WPOPOPFNZYPKAV-UHFFFAOYSA-N | |
| cyclobutylmethanol | |
| 78130 | |
| ≥98.0% (GC) | |
| Cyclobutanemethanol |
| Colorless | |
| C5H10O | |
| 5 g | |
| cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465 | |
| C1CC(C1)CO | |
| 86.134 | |
| 86.13 | |
| Liquid |
Safety and Handling
TSCA : No