missing translation for 'onlineSavingsMsg'
Learn More

Cyclobenzaprine Hydrochloride 98.0+%, TCI America™

Catalog No. C31571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31571G 1 g
C31575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31571G Supplier TCI America Supplier No. C31571G

Chemical Identifiers

CAS 6202-23-9
Molecular Formula C20H22ClN
Molecular Weight (g/mol) 311.85
MDL Number MFCD00079039
InChI Key VXEAYBOGHINOKW-UHFFFAOYSA-N
Synonym 5-(3-Dimethylaminopropylidene)dibenzo[a,e]cycloheptatriene Hydrochloride
PubChem CID 22576
ChEBI CHEBI:3997
IUPAC Name hydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}propyl)amine chloride
SMILES [H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2C=CC2=CC=CC=C12

Specifications

Melting Point 217°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 311.85
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cyclobenzaprine Hydrochloride
RTECSNumber HP0875000
TSCA No