missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Cyclobenzaprine hydrochloride
Chemical Identifiers
| CAS | 6202-23-9 |
|---|---|
| Chemical Name or Material | Cyclobenzaprine hydrochloride |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC466250010
|
Thermo Scientific™
466250010 |
1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC466250050
|
Thermo Scientific™
466250050 |
5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 6202-23-9 | |
| Cyclobenzaprine hydrochloride |
Specifications
| 6202-23-9 | |
| Solubility aq. soln.: soluble. Other solubilities: soluble in methanol, ethanol | |
| 216.0°C to 218.0°C | |
| Cyclobenzaprine hydrochloride |
Safety and Handling
GHS H Statement Toxic if swallowed.
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Store locked up.
GHS Signal Word: Danger
EINECSNumber : 228-264-4
RUO – Research Use Only