missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Cyanocobalamin, 96%
CAS: 68-19-9 | C63H88CoN14O14P | 1355.39 g/mol
Supplier: Thermo Scientific Chemicals 405920050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cyanocobalamin | |
| 68-19-9 | |
| Red | |
| Authentic | |
| Glass bottle | |
| MFCD00151092 | |
| 15, 10212 | |
| Solubility in water: soluble | |
| [C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O | |
| 1355.39 | |
| 1355.38 | |
| Crystalline Powder or Crystals |
| 96% | |
| 3.15 to 3.45 (A361/A(547 to 559)), 1.7 to 1.9 (A361/A278) | |
| 12% max. (20mg, 105°C, 2 hrs) (vacuum) | |
| 96% | |
| C63H88CoN14O14P | |
| 5 g | |
| vitamin b12 | |
| YUWVGNQGDAPKCX-BVWPOUIRNA-L | |
| cobalt(2+);[(2R,3S,4R,5S)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] [(2R)-1-[3-[(1R,2R,3R,5Z,7S,10Z,12S,13S,15Z,17S,18S,19R)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-octamethyl- | |
| 129893524 | |
| 96% |
Chemical Identifiers
| 68-19-9 | |
| 1355.39 | |
| YUWVGNQGDAPKCX-BVWPOUIRNA-L | |
| 129893524 | |
| [C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O |
| C63H88CoN14O14P | |
| MFCD00151092 | |
| vitamin b12 | |
| cobalt(2+);[(2R,3S,4R,5S)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] [(2R)-1-[3-[(1R,2R,3R,5Z,7S,10Z,12S,13S,15Z,17S,18S,19R)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-octamethyl- |
Safety and Handling
EINECSNumber : 200-680-0
RTECSNumber : GG3750000
TSCA : TSCA
RUO – Research Use Only