missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cyanocobalamin, 96%

Catalog No. p-95328
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405925000 500 mg Glass bottle
AC405920010 1 g Glass bottle
AC405920050 5 g Glass bottle
AC405920250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC405925000 Supplier Thermo Scientific Chemicals Supplier No. 405925000

CAS: 68-19-9 | C63H88CoN14O14P | 1355.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68-19-9
Molecular Formula C63H88CoN14O14P
Molecular Weight (g/mol) 1355.39
MDL Number MFCD00151092
InChI Key YUWVGNQGDAPKCX-BVWPOUIRNA-L
Synonym vitamin b12
PubChem CID 129893524
IUPAC Name cobalt(2+);[(2R,3S,4R,5S)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] [(2R)-1-[3-[(1R,2R,3R,5Z,7S,10Z,12S,13S,15Z,17S,18S,19R)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-octamethyl-
SMILES [C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O

Specifications

Absorbance 3.15 to 3.45 (A361/A(547 to 559)), 1.7 to 1.9 (A361/A278)
Color Red
Loss on Drying 12% max. (20mg, 105°C, 2 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 500 mg
Merck Index 15, 10212
Solubility Information Solubility in water: soluble
Formula Weight 1355.38
Percent Purity 96%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Cyanocobalamin, 96%
Show More Show Less
EINECSNumber 200-680-0
RTECSNumber GG3750000
TSCA TSCA

RUO – Research Use Only