missing translation for 'onlineSavingsMsg'
Learn More

Cyanide Standard, 25 ppm CN-, Ricca Chemical

Catalog No. R2540000100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R2540000100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R2540000100 Supplier Ricca Chemical Company Supplier No. R2540000100B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Accurate, ready-to-use stock standard solution for cyanide analysis, formulated to support consistent calibration and reliable ion-selective electrode performance.

Chemical Identifiers

CAS 7732-18-5

Specifications

Packaging Amber Poly Bottle
Odor Almond-like
Physical Form Liquid
Color Colorless
pH Alkaline
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Infinitely soluble in water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Cyanide Standard
Show More Show Less