missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cy-vBRIDP(regR) 90.0+%, TCI America™
Supplier: TCI America C29811G
Specifications
| Cy-vBRIDP(regR) | |
| 130°C | |
| C27H35P | |
| 1 g | |
| YMSBPYCREGBACF-UHFFFAOYSA-N | |
| dicyclohexyl(1,1-diphenylprop-1-en-2-yl)phosphane | |
| 12010824 | |
| ≥90.0% (HPLC) |
| 384842-24-4 | |
| White-Yellow | |
| MFCD10565627 | |
| Dicyclohexyl(1,1-diphenyl-1-propen-2-yl)phosphine | |
| CC(=C(C1=CC=CC=C1)C2=CC=CC=C2)P(C3CCCCC3)C4CCCCC4 | |
| 390.551 | |
| 390.55 | |
| Crystalline Powder |
Chemical Identifiers
| 384842-24-4 | |
| 390.551 | |
| YMSBPYCREGBACF-UHFFFAOYSA-N | |
| 12010824 | |
| CC(=C(C1=CC=CC=C1)C2=CC=CC=C2)P(C3CCCCC3)C4CCCCC4 |
| C27H35P | |
| MFCD10565627 | |
| Dicyclohexyl(1,1-diphenyl-1-propen-2-yl)phosphine | |
| dicyclohexyl(1,1-diphenylprop-1-en-2-yl)phosphane |
Safety and Handling
TSCA : No