missing translation for 'onlineSavingsMsg'
Learn More

Cy-vBRIDP(regR) 90.0+%, TCI America™

Catalog No. p-7125760
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2981200MG 200 mg
C29811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2981200MG Supplier TCI America Supplier No. C2981200MG
Only null left

Chemical Identifiers

CAS 384842-24-4
Molecular Formula C27H35P
Molecular Weight (g/mol) 390.551
MDL Number MFCD10565627
InChI Key YMSBPYCREGBACF-UHFFFAOYSA-N
Synonym Dicyclohexyl(1,1-diphenyl-1-propen-2-yl)phosphine
PubChem CID 12010824
IUPAC Name dicyclohexyl(1,1-diphenylprop-1-en-2-yl)phosphane
SMILES CC(=C(C1=CC=CC=C1)C2=CC=CC=C2)P(C3CCCCC3)C4CCCCC4

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 200 mg
Formula Weight 390.55
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cy-vBRIDP(regR)