missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Custom TaqMan™ Gene Expression Assay, FAM
GREENER_CHOICE

Catalog No. p-7180320
Encompass
Change view
Click to view available options
Quantity:
L (2900 reactions), made to order
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4332079 L (2900 reactions), made to order
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4332079 Supplier Applied Biosystems™ Supplier No. 4332079
Only null left

Applied Biosystems Custom TaqMan Gene Expression assays are the custom version of our predesigned TaqMan assays for quantitative real-time PCR.

Applied Biosystems Custom TaqMan Gene Expression assays are the custom version of our predesigned TaqMan assays for quantitative real-time PCR. The Custom TaqMan Gene Expression assays consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM) on the 5’ end and a minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3’ end.

Features include:
Easy to use—just add cDNA and master mix and run the qPCR—no melt curves required
Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
Accurate—identify small fold-changes with high accuracy of quantitation
Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
Sequences provided with every Custom TaqMan Gene Expression Assay you design and order

Approximate ship time
Made to order: 4–6 days in North America, 4–8 days in Europe

Pre-formulated assay
• 2 unlabeled PCR primers (900 nM each final 1X concentration)
• 1 FAM dye-labeled TaqMan MGB probe (250 nM final 1X concentration)

TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Recommended master mix: TaqMan Fast Advanced Master Mix

See also: Custom Plus TaqMan RNA Assays, designed using the full bioinformatic power of the TaqMan Assay design pipeline.

Specifications

Concentration 60X
Content And Storage 1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
Product Line TaqMan
Product Type Custom TaqMan Gene Expression
Quantity L (2900 reactions), made to order
Shipping Condition Room Temperature
No. of Reactions 2900 reactions, made to order
What is the bookmarking feature in the TaqMan Genotyper Software, and how would I use it?

Bookmarking is a unique feature in TaqMan Genotyper Software that allows you to tag a data point or well while reviewing results in a Study. For example, in reviewing a cluster plot for an assay, a data point is observed to be somewhat between clusters. You can set a bookmark for this data point to denote this well for further investigation. The bookmark persists between the Results workspace and Quality Control workspace, so you can easily identify the data point in a cluster plot, experiment plate view, or on the samples tab. Bookmarks are cleared upon exit from a Study or exit from the application.

I am getting the message: "An error has occurred. See the log file C:\ProgramFiles\Applied Biosystems\TaqMan Genotyper\config\eclipse\1363113099385.log." How can I fix this?

1.Go to the Start button, then Programs, then TaqMan Genotyper Software
2.Right-click on the program and choose “Run as Administrator”
3.If that does not work, go back to the same menu and choose “Properties”
4.Choose the “Compatibility” tab, and check “Run this program as administrator”
5.Click “Apply”
6.You may+C69 have to restart the computer for the settings to apply

Can I check the end product of a TaqMan Gene Expression Assay by running an agarose gel?

Yes, you can check the end product on an agarose gel. If you use the TaqMan Universal PCR Master Mix (including AmpErase UNG, Cat. No. 4304437), we recommend running the gel immediately after the PCR, since the trace amount of AmpErase UNG may digest the end product that contains double-stranded DNA with dUTP incorporated. The average length of amplicon ranges from 50-150 bp. The exact amplicon length is indicated on the detailed page of each assay on our website. You can get to that page by clicking on the Assay ID; the amplicon information is listed in the section "Assay Details".

Can I set up and store the TaqMan Fast Universal PCR Master Mix at room temperature if the instrument is in use?

For optimal results, we recommend to run the reaction plate right after setting up the reaction. If a reaction plate cannot be run within 2 hours of completing the reaction setup, then freeze/refrigerate the reaction plate until it can be loaded and run on the fast PCR instrument platform.

Can I run a reaction made with the standard TaqMan Universal PCR Master Mix in fast thermal cycling mode?

No, we do not recommend this approach. Since the standard TaqMan Universal PCR Master Mix is not optimized for running with fast thermal cycling, assay results will be significantly compromised.

Is the TaqMan Fast Universal PCR Master Mix offered with AmpErase UNG?

The TaqMan Fast Universal PCR Master Mix is provided without any UNG and you should add in AmpEraseUNG to reactions separately, with the addition of an initial 2 minute cycling step at 50º C. If you prefer to have a master mix with the UNG pre-added, please use TaqMan Fast Advanced Master Mix.

Can I compare performance of assays between the TaqMan Fast Universal PCR Master Mix compare and TaqMan Universal PCR Master Mix?

In general, assays have been found to perform comparably with these reagents, but they may not provide exactly identical results.

What are the storage conditions and the shelf life of the TaqMan Fast Universal PCR Master Mix?

We recommend a storage temperature of 4C and the shelf life is 9 months from the manufacturing date.

What is the recommended input amount of template for use with the TaqMan Fast Universal PCR Master Mix?

We recommend adding 10 to 100 ng of RNA converted to cDNA as your template.

Is there a TaqMan PCR Core Reagents kit for the Fast Real-Time PCR System?

No. The only Fast reagents that we offer are the TaqMan Fast Universal PCR Master Mix (2X) for single plex use, and Fast Advanced Master Mix for greater sensitivity and multiplexing use.

Can I use TaqMan Fast Universal PCR Master Mix for both one-step and two-step RT-PCR?

TaqMan Fast Universal PCR Master Mix is recommended for two-step RT-PCR, but for one-step reactions, we recommend using TaqMan Fast Virus 1-Step Master Mix as the preferred reagent.

What component in the TaqMan Fast Universal PCR Master Mix (P/N 4352042) allows rapid amplification results?

The hot-start DNA polymerase enzyme system in the TaqMan Fast Universal PCR Master Mix does not require an activation step, and can be activated with significantly shorter hold times for the thermal cycling stages of PCR, thereby minimizing PCR run time. For more information on using this reagent, please consult the TaqMan Fast Universal PCR Master Mix (2X) protocol.

What is the lowest copy number that can be used with the TaqMan Fast Universal PCR Master Mix (P/N 4352042)?

The fast master mix has demonstrated detection down to 10 copies of the RNase P gene with genomic DNA. Of course, detection limits may vary depending on factors such as assay design and sample preparation.

Which TaqMan assay types can be used when using the with TaqMan Fast Universal PCR Master Mix (P/N 4352042)?

The following research assays are supported using the TaqMan Fast Universal and/or Advanced Fast PCR Master Mix: TaqMan Gene Expression Assays, Custom TaqMan Gene Expression Assays, Custom primers and probes designed from Primer Express software-designed quantitative assays.

How can I obtain the primer and probe sequences for a custom TaqMan Gene Expression Assay that I designed using the Custom Assay Design Tool?

The primer and probe sequences for your custom TaqMan Gene Expression Assay can be found in the Assay Information File (AIF) which can be downloaded here. Once there, select the option "TaqMan™ or Absolute Q™ Assay" and, on the next page, enter the Sales Order number and the Rack/Plate ID for the respective assay (these numbers can be found on the assay packaging near the barcode), and submit. From there you can download a folder including a txt-file with all sequences.
Please note that the sequences are only accessible once the assays have been received.

What are the concentrations of the pre-formulated assay mix for the Custom TaqMan Gene Expression Assay, FAM?
How do I select a TaqMan Gene Expression Assay for an endogenous control in relative quantitative PCR (qPCR)?

An endogenous control gene is expressed at a consistent level in all studied conditions. It is impossible to predict exactly how any gene will behave under a given range of conditions. The best way of selecting the most appropriate control gene for a relative qPCR experiment is to select some candidate genes and determine their expression levels across the range of experimental conditions and treatments. The genes most stably expressed across these conditions will be the most appropriate controls. See the following links on selection and usage. Check the TaqMan Endogenous Control Assay Selector Tool here.

How can I reorder a Custom TaqMan Assay that I ordered previously?

To reorder the same Custom TaqMan Assay, you can follow these steps:
- Select an appropriate catalog number, considering the size of the assay that you want. You can find the different catalog numbers for Custom TaqMan Assays on their respective product pages on thermofisher.com.
- Once you have identified the necessary catalog number, go to www.thermofisher.com and click on "Quick Order", in the top right corner of the screen.
- In the following page, paste your selected catalog number in the "Catalog Number" field and paste the assay ID from your previous order in the "Assay ID" field.
- Click "Add to cart" and then in the following screen click "Proceed to check out".

Can I perform a one-step RT-PCR reaction using TaqMan Gene Expression Assays?

While TaqMan Gene Expression Assays can be used to perform a one-step RT-PCR reaction, these assays have not been optimized for this application. TaqMan Gene Expression Assays have been designed, tested, and optimized using Applied Biosystems universal conditions for a two-step RT-PCR reaction.

When preparing the TaqMan or Custom TaqMan Gene Expression Assays, what is the recommended reaction volume?

When preparing the TaqMan or Custom TaqMan Gene Expression Assays, the recommended and supported volume is 20-50 µL for a 96-well standard plate set-up; 10-20 µL for a 96-well fast plate set-up and 5-10 µL for a 384-well set-up. For more information, please consult the TaqMan Gene Expression Assays and Custom TaqMan Gene Expression Assays protocols.

How much cDNA is recommended per TaqMan or Custom TaqMan Gene Expression reaction?

For optimal performance of the TaqMan and Custom TaqMan Gene Expression Assays, use 1 to 100 ng of cDNA per 50 µL total reaction volume. For more information, please consult the TaqMan Gene Expression Assay protocol and Custom TaqMan Gene Expression Assay protocol.

What are the storage conditions for the TaqMan and Custom TaqMan Gene Expression and SNP Genotyping Assays?

After shipment, the assay mixes should be stored at -15 C to -20 C to maximize stability. Freeze/thaws should be kept to a minimum (a maximum of 10 freeze-thaw cycles is recommended). Keep all TaqMan and Custom TaqMan Gene Expression and SNP Genotyping Assays protected from direct exposure to light. Excessive exposure to light may affect the fluorescent probes. If desired, the Assay Mix may be diluted with TE buffer or DNase-free Water. It is recommended to make working stocks from the stock solutions provided in these kits.

Can I use SYBR Green I dye in conjunction with the TaqMan Gene Expression Assays or Custom TaqMan Gene Expression Assays?

No, SYBR Green dye I is not compatible with the TaqMan Gene Expression Assays or Custom TaqMan Gene Expression Assays. TaqMan Gene Expression Assays or Custom TaqMan Gene Expression Assays contain TaqMan MGB probes combined with primers at non-limiting concentrations. The probes already have detection dyes conjugated to them, and SYBR Green is not needed. For more information, please consult the protocol of TaqMan Gene Expression Assays and Custom TaqMan Gene Expression Assays.

I seem to have misplaced the CD for my custom TaqMan assay I recently ordered. How can I get the details?

The details for custom TaqMan assays can be downloaded at: https://www.thermofisher.com/order/taqman-files. Select the option "TaqMan Assays" and, in the screen that follows, indicate the sales order number and the Rack ID of your assay.
Both these numbers can be found on the assay package, next to the barcode.

What is the difference between TaqMan Gene Expression Assays and Custom TaqMan Gene Expression Assays?

TaqMan Gene Expression Assays are pre-designed, quantitative, human gene expression assays based on the TaqMan probe-based chemistry. The assays are formulated into a 20X mix.

Custom TaqMan Gene Expression Assays or Custom Plus TaqMan RNA Assays are designed based on the target sequences submitted by the customer, then synthesized, formulated into 20X concentration, and delivered. The target sequence information can be submitted by the customer through the online Custom Assay Design Tool. The integrity of the sequence information must be verified before the design process. When you submit a sequence, you have an option to let us run the bioinformatic evaluation of the target sequence to verify the integrity (Custom Plus) or you can run the bioinformatic evaluation yourself. This integrity verification can be accomplished through BLAST analysis or other methods of verifying sequence integrity. See the Custom TaqMan Assays Design and Ordering Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/cms_042307.pdf) for more information.

Can TaqMan Gene Expression Assays be used in a multiplex reaction?

Yes. For duplexing, we have VIC-labeled probes with primer limited concentrations (VIC PL) available from our pre-designed assay selection. The primer limited assay is used to detect the most abundant gene. For guidance on multiplexing, see the TaqMan Multiplex PCR Optimization User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/taqman_optimization_man.pdf).

What are the components of a TaqMan Gene Expression Assay?

Each TaqMan Gene Expression Assay consists of two unlabeled PCR primers and a FAM or VIC dye-labeled TaqMan MGB probe (with non-fluorescent quencher). All the components are combined into a 20X formulation. Purchase of a TaqMan Gene Expression Assay does not provide TaqMan PCR Master Mix.

Why is there is a noisy signal above the threshold for my TaqMan Gene Expression Assay?

Please review the following possible causes and solutions:
-The sample evaporated. Check the seal of the adhesive film for leaks.
- The well is empty because of inaccurate pipetting. Check the calibration of the pipettes, and pipette at least 5 µL of sample.
- The well is assigned a sample or target in the plate document or experiment, but the well is empty. Ensure that the plate document or experiment is set up correctly. Then try excluding the well and reanalyzing the data.

There was a small ΔRn for samples in my TaqMan Gene Expression Assay?

A small ΔRn can mean that the PCR efficiency was poor. Ensure that the reagents were used at the correct concentration. Alternatively, the quantity of the cDNA may be low (a low copy number of the target). In this case, we recommend that you increase the quantity of the cDNA in the reaction.

Why did the Rn value shift during the early PCR cycles (cycles 0 to 5) for my TaqMan Gene Expression Assay?

Most likely fluorescence did not stabilize to the buffer conditions of the reaction mix.
Note: This condition does not affect PCR or the final results.
We recommend that you try the following:
- Reset the lower value of the baseline range.
- Use an automatic baseline.
- Use the relative threshold algorithm (Crt). See Introduction to Gene Expression Getting Started Guide (Pub. No. 4454239).

Why are the results for my TaqMan Gene Expression inconsistent (high standard deviation in the replicates, inconsistent data, or a variable Ct)?

Please review the following possible causes and solutions:
- The reagents were not mixed properly. We suggest increasing the length of time that you mix the reagents and confirming your mixing process by running a replicate assay.
- Pipetting was inaccurate. We recommend that you check the pipette calibration, and pipette at least 5 µL of sample to prepare the reaction mix.
- The threshold was not set correctly. Set the threshold above the noise level and where the replicates are tightest. Please see your real‑time PCR system user documentation for more information about setting the threshold.
- There was a low concentration of the target of interest. Rerun the assay with more cDNA template.

Why do the endogenous control Ct values vary and/or do not normalize the sample well in my TaqMan Gene Expression Assay?

Please review the following possible causes and solutions:
- The endogenous control is not consistently expressed across the samples. Please ensure that the endogenous control is consistently expressed in your sample type. - The sample concentrations vary. We recommend that you quantify and normalize the PCR samples before running the assay.
- Pipetting was inaccurate. We recommend that you check the pipette calibration, and pipet at least 5 µL of sample to prepare the reaction mix.

Why does the no-template control (NTC) for my TaqMan Gene Expression Assay show amplification?

Most likely the reagents are contaminated with gDNA, amplicon or plasmid clones. We recommend that you clean your workspace and equipment, then rerun the assay using new reagents. We also recommend that you run no-RT controls to rule out genomic DNA contamination, and treat the sample with DNase.

Why is the Rn in the Rn vs. Cycle plot so high for my TaqMan Gene Expression Assay?

Most likely the ROX dye was not set as the passive reference. Set ROX dye as the passive reference, then reanalyze the data.

Why is the multicomponent signal for ROX dye not flat for my TaqMan Gene Expression Assay?

Most likely incorrect dyes were selected for each target. We suggest checking the dyes selected for each target, then reanalyzing the data.

Why was there a simultaneous increase in fluorescence from both the passive reference dye (ROX dye) and the reporter dyes for my TaqMan Gene Expression Assay?

The sample evaporated. We recommend that you check the seal of the adhesive film for leaks before running the plate on the real-time PCR instrument.

Why was there a decrease in ROX dye fluorescence (passive reference dye) in my TaqMan Gene Expression Assay?

It is possible that there was precipitation in the buffers. Before preapring reactions, we recommend that you mix the Master Mix thoroughly to produce a homogenous solution.
It as also possible that the reagents have degraded. Ensure that the kits and reagents have been stored according to the instructions on the packaging and that they have not expired.

Why does the amplification curve show no amplification of the sample (Ct=40) in the target TaqMan Gene Expression Assay?

Please review the following possible causes and solutions:
- The gene is not expressed in the sample. First, confirm that the gene is expressed in the sample type or tissue type at ncbi.nlm.nih.gov/unigene. If the gene is expressed, confirm the results by rerunning the sample using the same assay and/or rerunning the experiment using more of the sample. Avoid preparing PCR reaction mixes with more than 20% reverse transcription reaction. You can also run the experiment using an alternative assay, if available, that detects a different transcript or more than one transcript from the same gene.
Note: If the recommended actions do not resolve the problem, the result may be correct.

- The sample does not have enough copies of the target RNA. To confirm the results, we suggest rerunning the sample using the same assay and/or rerunning the assay using more of the sample. Avoid PCR reaction mix with more than 20% from the reverse transcription reaction.
Note: If the recommended actions do not resolve the problem, the result may be correct.

- One or more of the reaction components was not added. Please check your pipetting equipment and/or technique.

- Incorrect dyes were selected for each target. We recommend checking the dyes selected for each target, then reanalyze the data.

Why does the amplification curve show samples targeted by the same TaqMan Gene Expression Assay that have differently shaped curves?

Please review the following possible causes and solutions:
- The baseline was set improperly. See your real‑time PCR system user guide for procedures on setting the baseline. We recommend switching from an automatic baseline to a manual baseline (or vice versa) and/or increasing the upper or lower value of the baseline range.
- The sample quality was poor. We recommend performing a quality check on the sample, then re-extracting the sample if needed.
- There were different concentrations caused my imprecise pipetting. Please follow accurate pipetting practices.
- The reagents or equipment are contaminated. Please ensure that your workspace and equipment are cleaned properly.

Why does the multicomponent plot show low levels of ROX dye (passive reference dye) in the Taqman Gene Expression Assay?

Most likely little or no MasterMix is present in the reaction due to inaccurate pipetting. Please follow accurate pipetting practices when setting up reactions.

Why does the amplification curve show an abnormal plot and/or low ΔRn values for samples in my TaqMan Gene Expression Assay?

Please review the following possible causes and solutions:
- The baseline was set too high and some samples have Ct values lower than the baseline stop value. We recommend that you switch from manual to automatic baselining, or move the baseline stop value to a lower Ct. The baseline stop value should be set to a Ct 2 cycles before the amplification curve crosses the threshold. Please see your real‑time PCR system user guide for procedures on setting the baseline.
-No baseline can be set because the amplification signal is detected too early in the PCR cycles. Diluting the sample can increase the Ct value.

Why did amplification occur in the no-RT control for my TaqMan Gene Expression Assay?

Please review the following possible causes and solutions:
-Genomic DNA (gDNA) contamination occured. We recommend that you improve sample extraction methods and use DNase to ensure minimal gDNA contamination of the RNA.
For custom assays, we recommend that you design an assay that spans an exon-exon boundary. Please see the Custom TaqMan Assays Design and Ordering Guide (Pub. No. 4367671).
- The cDNA template or amplicon is contaminated. In this case, please follow established PCR good laboratory practices.

Why is the Ct value for samples in my TaqMan Gene Expression Assay lower than expected?

Please review the following possible causes and solutions:
- Contamination occurred. We recommend that you run a no-RT control to confirm that there was genomic DNA (gDNA) contamination. We also recommend the use DNase to ensure minimal gDNA contamination of the RNA.
For custom assays, we recommend that you design an assay that spans an exon-exon boundary. Please see the Custom TaqMan Assays Design and Ordering Guide (Pub. No. 4367671).
- Too much cDNA template was added to the reaction. We recommend that you quantitate the RNA before the reverse transcription (RT) reaction. After the RT reaction, adjust the concentration of cDNA before adding it to the reaction.
- The cDNA template or the amplicon is contaminated. Follow established PCR good laboratory practices.

Why does the amplification curve show no amplification of the sample (Ct=40) across all TaqMan Gene Expression Assays or in an unusually large number of assays?

Please review the following possible causes and solutions:
- One or more of the reaction components was not added. Ensure that the cDNA, the assay and the Master Mix were added to the reaction plate. The passive reference fails if the Master Mix is missing.

- Incorrect dyes were selected for each target. Check the dyes selected for each target, then reanalyze the data.

- The annealing temperature was too high for the primers and/or probe. Ensure that the correct annealing and extension temperatures are set, and that the real-time PCR instrument is calibrated and maintained regularly.

- Inappropriate reaction conditions were used. Ensure that the properties and the thermal protocol are correct, then troubleshoot the real-time PCR optimization.
- The template is degraded. Determine the quality of the template, then rerun the assay with fresh template if needed. We recommend that you use use RNase-free reagents and an RNase inhibitor.

- Inhibitors are present in the reaction. To check for inhibitors, we recommend that you run a serial dilution of your sample with an expressing assay (for example, an endogenous control). If an inhibitor is present, the high concentration samples yield higher-than-expected Ct values because the samples are not diluted.

- The baseline and/or threshold was improperly set. See your real-time PCR system user guide for procedures on setting the baseline and threshold. Some possible solutions to this issue are switching from an automatic baseline to a manual baseline (or vice versa), and/or lowering the threshold value to fall within the appropriate range.

- The reverse transcription failed. We recommend checking the RNA integrity and concentration, checking for RNase activity, following the recommended thermal profile, and/or repeating the reverse transcription using new reagents.

- (Custom TaqManGene Expression Assays only) The design or synthesis of the custom assay failed. Ensure that the sequence you submitted is correct. We also recomend that you check for an alternative trascript or splice variant.

- (Custom TaqManGene Expression Assays only) The assay is designed in a variable regions of the gene transccript. Ensure that the location that is targeted by the assay is not within the 5' untranslated region (UTR), which can be highly variable between transcripts. If the assay is designed within the 5' UTR, we recommend that you select a different assay that is within the coding region of the transcript. Otherwise, select an assay for an alternative transcriptor splice variant.

Why does the amplification curve show weak amplification for the samples in my Custom Gene Expression Assay?

The sequence provided for the assay design may contain mismatches with sample sequences. We recommend that you perform bioinformatics analysis for assay design.
For more information, see the Custom TaqMan Assays Design and Ordering Guide (Pub.No. 4367671).

Why does the amplification curve show a rising baseline for samples in my TaqMan Gene Expression Assay?

There may be interaction between the primer and probe. We recommend that you adjust the threshold manually, or select another assay from the same gene, if available.

How do I get the assay information file for my Custom or Custom Plus TaqMan RNA assay?

For orders placed after March 2016, assay information files can be accessed from this page: www.thermofisher.com/taqmanfiles

How many reactions can I get out of the Custom and Custom Plus TaqMan Gene Expression assays?

The number of reactions will depend on your final reaction volume. The Custom and Custom Plus TaqMan RNA assays are supplied with the following volumes:
Small: 360 µL (20X)
Medium: 750 µL (20X)
Large: 976 µL (60X)

Assuming a 20 µL final reaction volume, the small size would provide 360 reactions, the medium size would provide 750 reactions, and the large size would provide 2,900 reactions.

What volume will I receive with the Custom and Custom Plus TaqMan RNA assays for the different scales?

The Custom and Custom Plus TaqMan RNA assays are supplied with the following volumes:
Small: 360 µL
Medium: 750 µL
Large: 976 µL

Please note that the large scale is supplied in a 60X format, while the small and medium scales are supplied in 20X format.

What formats are available for Custom and Custom Plus TaqMan RNA assays?

Custom and Custom Plus TaqMan RNA assays can be ordered online with either FAM, VIC, or VIC primer limited.

Which reporter dyes and quenchers are available for Custom TaqMan RNA assays and Custom Plus TaqMan RNA assays?

Custom and Custom Plus TaqMan RNA assays can be ordered online with either FAM or VIC dye, with a MGB-NFQ quencher.

Do you sell human endogenous control assays for normalizing sample loading in gene expression studies?

Yes, we offer two options for obtaining endogenous control assays for gene expression studies:

The first option is our specially designed TaqMan Endogenous Controls products, which are a collection of probe and primer sets that are optimized, pre-formulated, and ready-to-use specifically as control assays to save on time. Endogenous Control formulations are available as primer limited and contain probes labeled with FAM or VIC reporter dyes. This allows multiplexing of TaqMan Endogenous Controls with TaqMan target gene primer and probe sets, provided that the control gene is more abundantly expressed than the target gene.

Another option is to simply order a general TaqMan Gene Expression Assay for genes that are commonly used as endogenous controls, such as 18S rRNA, GAPDH, beta-actin, etc. The TaqMan Gene Expression Assays are biologically informative, pre-formulated assays for rapid, reliable detection and quantification of human mRNA transcripts. They come in a wide variety of reaction sizes and formats, and may be ordered with either a FAM or VIC reporter dye label with MGB quenchers. You can also specify that a VIC-labeled probe should be Primer Limited (PL) for multiplexing.


For Research Use Only. Not for use in diagnostic procedures.