missing translation for 'onlineSavingsMsg'
Learn More

Cuprous Chloride Reagent, Acid, Ricca Chemical

Catalog No. 250016
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
250016 500 mL Amber Glass Bottle
250032 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 250016 Supplier Ricca Chemical Company Supplier No. 250016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Analysis of Carbon Monoxide Absorbent For orsat Gas Analysis

Chemical Identifiers

CAS 7758-89-6 , 7732-18-5 , 7647-01-0
Molecular Formula ClCu
Molecular Weight (g/mol) 99.00
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
PubChem CID 62652  
ChEBI CHEBI:53472
IUPAC Name λ¹-copper(1+) chloride
SMILES [Cl-].[Cu+]

Specifications

CAS Min % 4.25, 54.0, 40.12
CAS Max % 4.43, 56.0, 41.76
Packaging Amber Glass Bottle
Quantity 500 mL
Chemical Name or Material Cuprous Chloride Reagent