missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupriethylenediamine, 1.0 M, Solution, Spectrum™ Chemical
C1449, 14552-35-3, C4H18N4O2Cu
$421.46 - $1211.82
Chemical Identifiers
| CAS | 13426-91-0 |
|---|---|
| Molecular Formula | C4H16CuN4 |
| Molecular Weight (g/mol) | 183.75 |
| InChI Key | ATSGLBOJGVTHHC-UHFFFAOYSA-N |
| IUPAC Name | copper(2+) bis(ethane-1,2-diamine) |
| SMILES | [Cu++].NCCN.NCCN |
Description
Spectrum™ Chemical Cupriethylenediamine, 1.0 M, SolutionChemical Identifiers
| 13426-91-0 | |
| 183.75 | |
| copper(2+) bis(ethane-1,2-diamine) |
| C4H16CuN4 | |
| ATSGLBOJGVTHHC-UHFFFAOYSA-N | |
| [Cu++].NCCN.NCCN |
Specifications
| Purple | |
| 22.5%,77.5% | |
| UN1761 | |
| ATSGLBOJGVTHHC-UHFFFAOYSA-N | |
| copper(2+) bis(ethane-1,2-diamine) | |
| Chemical Solution | |
| 12 | |
| Cupriethylenediamine |
| 13426-91-0 , 7732-18-5 | |
| C4H16CuN4 | |
| 1.0 M | |
| [Cu++].NCCN.NCCN | |
| 183.75 | |
| Amber Glass Bottle | |
| 1 L |